Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACACCCCCCTCCCCCACCTCTT[C/T]CCCAATATCTCAGATCCCAAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604590 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGR2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCGR2B - Fc fragment of IgG receptor IIb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002273.2 | Intron | NP_001002273.1 | ||||
NM_001002274.2 | Intron | NP_001002274.1 | ||||
NM_001002275.2 | Intron | NP_001002275.1 | ||||
NM_001190828.1 | Intron | NP_001177757.1 | ||||
NM_004001.4 | Intron | NP_003992.3 | ||||
XM_011509292.2 | Intron | XP_011507594.1 | ||||
XM_017000669.1 | Intron | XP_016856158.1 | ||||
XM_017000670.1 | Intron | XP_016856159.1 | ||||
XM_017000671.1 | Intron | XP_016856160.1 | ||||
XM_017000672.1 | Intron | XP_016856161.1 |
RPL31P11 - ribosomal protein L31 pseudogene 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |