Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAATTTAATCTGAATTGCTTGCA[A/G]ATACTGCTAATTTTCTTTTTCTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609424 | ||||||||||||||||||||
Literature Links: |
RC3H1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RC3H1 - ring finger and CCCH-type domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300850.1 | 3715 | UTR 3 | NP_001287779.1 | |||
NM_001300851.1 | 3715 | UTR 3 | NP_001287780.1 | |||
NM_001300852.1 | 3715 | UTR 3 | NP_001287781.1 | |||
NM_172071.3 | 3715 | UTR 3 | NP_742068.1 | |||
XM_005244921.3 | 3715 | UTR 3 | XP_005244978.1 | |||
XM_011509231.2 | 3715 | UTR 3 | XP_011507533.1 |