Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGAACATGGAGTGAGGAAGAG[A/G]TAGTGTTATAAATGACTCCAAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL20 - kelch like family member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014458.3 | Intron | NP_055273.2 | ||||
XM_005245093.3 | Intron | XP_005245150.1 | ||||
XM_011509420.2 | Intron | XP_011507722.1 | ||||
XM_017001052.1 | Intron | XP_016856541.1 | ||||
XM_017001053.1 | Intron | XP_016856542.1 | ||||
XM_017001054.1 | Intron | XP_016856543.1 |
Set Membership: |
HapMap |