Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTAGGGCTTCATTACATAAA[C/G]TTCACTGATTGCCCACATGGCTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605573 MIM: 609182 | ||||||||||||||||||||
Literature Links: |
HSD17B3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD17B3 - hydroxysteroid 17-beta dehydrogenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000197.1 | Intron | NP_000188.1 | ||||
XM_011518618.2 | Intron | XP_011516920.1 | ||||
XM_011518619.2 | Intron | XP_011516921.1 | ||||
XM_017014671.1 | Intron | XP_016870160.1 | ||||
XM_017014672.1 | Intron | XP_016870161.1 | ||||
XM_017014673.1 | Intron | XP_016870162.1 | ||||
XM_017014674.1 | Intron | XP_016870163.1 | ||||
XM_017014675.1 | Intron | XP_016870164.1 | ||||
XM_017014676.1 | Intron | XP_016870165.1 | ||||
XM_017014677.1 | Intron | XP_016870166.1 |
SLC35D2 - solute carrier family 35 member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286990.1 | Intron | NP_001273919.1 | ||||
NM_007001.2 | Intron | NP_008932.2 | ||||
XM_005251673.1 | Intron | XP_005251730.1 | ||||
XM_005251674.1 | Intron | XP_005251731.1 | ||||
XM_005251675.2 | Intron | XP_005251732.1 | ||||
XM_005251676.1 | Intron | XP_005251733.1 | ||||
XM_005251678.4 | Intron | XP_005251735.1 | ||||
XM_006716939.2 | Intron | XP_006717002.1 | ||||
XM_011518164.1 | Intron | XP_011516466.1 |