Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACACACAGTAGACTATTTCTACC[C/T]GCAATTCTCTTATTCTCTTTGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137166 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABRG1 - gamma-aminobutyric acid type A receptor gamma1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173536.3 | 6281 | UTR 3 | NP_775807.2 | |||
XM_017007990.1 | 6281 | UTR 3 | XP_016863479.1 |
Set Membership: |
HapMap |