Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACTGTATCCAGTCTAAGTAAGT[A/G]TAGCCCAACATGGACCAACAAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609811 | ||||||||||||||||||||
Literature Links: |
COX7B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX7B2 - cytochrome c oxidase subunit 7B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130902.2 | Intron | NP_570972.2 | ||||
XM_005248056.4 | Intron | XP_005248113.1 | ||||
XM_011513630.2 | Intron | XP_011511932.1 | ||||
XM_011513631.2 | Intron | XP_011511933.1 | ||||
XM_011513632.2 | Intron | XP_011511934.1 | ||||
XM_011513633.2 | Intron | XP_011511935.1 | ||||
XM_011513634.2 | Intron | XP_011511936.1 | ||||
XM_011513635.2 | Intron | XP_011511937.1 | ||||
XM_011513636.1 | Intron | XP_011511938.1 | ||||
XM_011513637.2 | Intron | XP_011511939.1 | ||||
XM_011513638.2 | Intron | XP_011511940.1 |