Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTAAGTATTTTAGTTTCCTGA[C/T]TGTGGGCATGTGAGTAAAGTCAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601192 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLA2G5 - phospholipase A2 group V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000929.2 | Intron | NP_000920.1 | ||||
XM_005245891.4 | Intron | XP_005245948.1 | ||||
XM_005245892.4 | Intron | XP_005245949.1 | ||||
XM_005245893.4 | Intron | XP_005245950.1 | ||||
XM_011541586.2 | Intron | XP_011539888.1 | ||||
XM_011541587.2 | Intron | XP_011539889.1 | ||||
XM_011541588.2 | Intron | XP_011539890.1 | ||||
XM_011541589.2 | Intron | XP_011539891.1 | ||||
XM_011541590.2 | Intron | XP_011539892.1 | ||||
XM_011541591.2 | Intron | XP_011539893.1 | ||||
XM_011541592.2 | Intron | XP_011539894.1 |
Set Membership: |
HapMap |