Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATGTATACACACATCTGAATCT[A/G]TTTCTATGCCTATGTACAGGTATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176878 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTPN4 - protein tyrosine phosphatase, non-receptor type 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002830.3 | Intron | NP_002821.1 | ||||
XM_011511560.2 | Intron | XP_011509862.1 | ||||
XM_011511562.2 | Intron | XP_011509864.1 | ||||
XM_011511563.2 | Intron | XP_011509865.1 | ||||
XM_017004596.1 | Intron | XP_016860085.1 | ||||
XM_017004597.1 | Intron | XP_016860086.1 | ||||
XM_017004598.1 | Intron | XP_016860087.1 | ||||
XM_017004599.1 | Intron | XP_016860088.1 | ||||
XM_017004600.1 | Intron | XP_016860089.1 | ||||
XM_017004601.1 | Intron | XP_016860090.1 | ||||
XM_017004602.1 | Intron | XP_016860091.1 | ||||
XM_017004603.1 | Intron | XP_016860092.1 | ||||
XM_017004604.1 | Intron | XP_016860093.1 | ||||
XM_017004605.1 | Intron | XP_016860094.1 | ||||
XM_017004606.1 | Intron | XP_016860095.1 | ||||
XM_017004607.1 | Intron | XP_016860096.1 | ||||
XM_017004608.1 | Intron | XP_016860097.1 |
Set Membership: |
HapMap |