Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCAAAAATGGAAAAATATGACA[A/T]TTCCATGCAAAGAAATGTATCTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603390 | ||||||||||||||||||||
Literature Links: |
PDE8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR41 - WD repeat domain 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018268.3 | 3983 | Intron | NP_060738.2 | |||
XM_005248551.4 | 3983 | Intron | XP_005248608.1 | |||
XM_005248552.3 | 3983 | Intron | XP_005248609.1 | |||
XM_011543505.2 | 3983 | Intron | XP_011541807.1 | |||
XM_011543507.2 | 3983 | Intron | XP_011541809.1 | |||
XM_017009606.1 | 3983 | Intron | XP_016865095.1 |