Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609073 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXW8 - F-box and WD repeat domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012174.1 | Intron | NP_036306.1 | ||||
NM_153348.2 | Intron | NP_699179.2 | ||||
XM_005253867.2 | Intron | XP_005253924.1 | ||||
XM_005253868.3 | Intron | XP_005253925.1 | ||||
XM_017019175.1 | Intron | XP_016874664.1 | ||||
XM_017019176.1 | Intron | XP_016874665.1 | ||||
XM_017019177.1 | Intron | XP_016874666.1 | ||||
XM_017019178.1 | Intron | XP_016874667.1 |