Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163731 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOS1 - nitric oxide synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000620.4 | 7366 | UTR 3 | NP_000611.1 | |||
NM_001204213.1 | 7366 | UTR 3 | NP_001191142.1 | |||
NM_001204214.1 | 7366 | UTR 3 | NP_001191143.1 | |||
NM_001204218.1 | 7366 | UTR 3 | NP_001191147.1 | |||
XM_011538398.2 | 7366 | UTR 3 | XP_011536700.1 | |||
XM_017019345.1 | 7366 | UTR 3 | XP_016874834.1 | |||
XM_017019346.1 | 7366 | UTR 3 | XP_016874835.1 | |||
XM_017019347.1 | 7366 | UTR 3 | XP_016874836.1 |
Set Membership: |
HapMap |