Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGATTGAGAACTTTCTATCTGCT[C/T]ACTAGACTCTAACTTCAAGTCTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616381 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZCCHC8 - zinc finger CCHC-type containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017612.4 | Intron | NP_060082.2 | ||||
XM_006719491.2 | Intron | XP_006719554.1 | ||||
XM_011538555.2 | Intron | XP_011536857.1 | ||||
XM_017019606.1 | Intron | XP_016875095.1 |
Set Membership: |
HapMap |