Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAACCCCACTGCTTACAATCCA[C/T]ATCTCATCAGATTCTCATGACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608225 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT14 - polypeptide N-acetylgalactosaminyltransferase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253826.1 | Intron | NP_001240755.1 | ||||
NM_001253827.1 | Intron | NP_001240756.1 | ||||
NM_024572.3 | Intron | NP_078848.2 | ||||
XM_011533105.1 | Intron | XP_011531407.1 | ||||
XM_017004906.1 | Intron | XP_016860395.1 | ||||
XM_017004907.1 | Intron | XP_016860396.1 | ||||
XM_017004908.1 | Intron | XP_016860397.1 |
Set Membership: |
HapMap |