Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGGCACTTAGCCTTCAATGA[C/G]TGGAGGCCCACTGGCAACATTTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605928 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARFIP1 - ADP ribosylation factor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FHDC1 - FH2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033393.2 | Intron | NP_203751.2 | ||||
XM_005263319.3 | Intron | XP_005263376.1 | ||||
XM_006714393.3 | Intron | XP_006714456.1 | ||||
XM_011532388.1 | Intron | XP_011530690.1 | ||||
XM_011532389.1 | Intron | XP_011530691.1 | ||||
XM_017008787.1 | Intron | XP_016864276.1 |