Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAGGACACAGACAAGTACCATG[A/T]GAAAGATGAATCCTCCTTTGCCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300127 | ||||||||||||||||||||
Literature Links: |
OPHN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OPHN1 - oligophrenin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002547.2 | Intron | NP_002538.1 | ||||
XM_005262270.1 | Intron | XP_005262327.1 | ||||
XM_006724653.2 | Intron | XP_006724716.1 | ||||
XM_011530961.1 | Intron | XP_011529263.1 | ||||
XM_017029555.1 | Intron | XP_016885044.1 |