Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTAGTAGTGCCCAGGAGACAGTT[A/C]TAAGCAAATTGTAAGCATTTTCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614726 | |||||||||||||||||||||||
Literature Links: |
TMEM165 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM165 - transmembrane protein 165 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018475.4 | Intron | NP_060945.2 | ||||
XM_011534394.2 | Intron | XP_011532696.1 | ||||
XM_017008412.1 | Intron | XP_016863901.1 |