Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAAACAAAGTATCAGAGGTCTC[A/G]GTAAGGGCAAAAACAGCTTTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611676 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDCL2 - phosducin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152401.2 | Intron | NP_689614.2 | ||||
XM_005265728.3 | Intron | XP_005265785.1 | ||||
XM_011534374.2 | Intron | XP_011532676.1 | ||||
XM_017007745.1 | Intron | XP_016863234.1 |
Set Membership: |
HapMap |