Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTAGGCCCAGCAAAGCGAACACT[C/T]TCCTGCAATTGCATTTCTATTCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605103 | ||||||||||||||||||||
Literature Links: |
NMU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NMU - neuromedin U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292045.1 | Intron | NP_001278974.1 | ||||
NM_001292046.1 | Intron | NP_001278975.1 | ||||
NM_006681.3 | Intron | NP_006672.1 | ||||
XM_011534367.2 | Intron | XP_011532669.1 | ||||
XM_011534368.2 | Intron | XP_011532670.1 |