Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAAGCAAAAGTGAAGGGCGA[G/T]AATTGCCCACAATCATTTTCTTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602867 MIM: 180661 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGFBP7 - insulin like growth factor binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253835.1 | Intron | NP_001240764.1 | ||||
NM_001553.2 | Intron | NP_001544.1 |
POLR2B - RNA polymerase II subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000938.2 | Intron | NP_000929.1 | ||||
NM_001303268.1 | Intron | NP_001290197.1 | ||||
NM_001303269.1 | Intron | NP_001290198.1 |
Set Membership: |
HapMap |