Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAGTTTATTTATTGTAGTTGTT[A/C]TTCGGCTCTGTGTGTGTGTGTGTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607449 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPFFR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NPFFR2 - neuropeptide FF receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144756.1 | Intron | NP_001138228.1 | ||||
NM_004885.2 | Intron | NP_004876.2 | ||||
NM_053036.2 | Intron | NP_444264.1 | ||||
XM_011531554.2 | Intron | XP_011529856.1 |
Set Membership: |
HapMap |