Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611339 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATG4C - autophagy related 4C cysteine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032852.3 | Intron | NP_116241.2 | ||||
NM_178221.2 | Intron | NP_835739.1 | ||||
XM_005271288.2 | Intron | XP_005271345.1 | ||||
XM_011542313.2 | Intron | XP_011540615.1 | ||||
XM_017002600.1 | Intron | XP_016858089.1 | ||||
XM_017002601.1 | Intron | XP_016858090.1 |
Set Membership: |
HapMap |