Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612257 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAST2 - microtubule associated serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319245.1 | Intron | NP_001306174.1 | ||||
NM_001324320.1 | Intron | NP_001311249.1 | ||||
NM_001324321.1 | Intron | NP_001311250.1 | ||||
NM_015112.2 | Intron | NP_055927.2 | ||||
XM_005270655.3 | Intron | XP_005270712.1 | ||||
XM_005270656.4 | Intron | XP_005270713.1 | ||||
XM_006710477.3 | Intron | XP_006710540.1 | ||||
XM_011541059.2 | Intron | XP_011539361.1 | ||||
XM_011541061.2 | Intron | XP_011539363.1 | ||||
XM_011541062.2 | Intron | XP_011539364.1 | ||||
XM_011541063.1 | Intron | XP_011539365.1 | ||||
XM_011541064.2 | Intron | XP_011539366.1 | ||||
XM_011541065.1 | Intron | XP_011539367.1 | ||||
XM_011541067.2 | Intron | XP_011539369.1 | ||||
XM_011541068.1 | Intron | XP_011539370.1 | ||||
XM_011541069.2 | Intron | XP_011539371.1 | ||||
XM_017000752.1 | Intron | XP_016856241.1 | ||||
XM_017000753.1 | Intron | XP_016856242.1 | ||||
XM_017000754.1 | Intron | XP_016856243.1 | ||||
XM_017000755.1 | Intron | XP_016856244.1 |
Set Membership: |
HapMap |