Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGACCTTTGCACTGACTATTC[C/G]CTTTGCCCGGTCCACTCTTGCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD2A - major facilitator superfamily domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136493.2 | Intron | NP_001129965.1 | ||||
NM_001287808.1 | Intron | NP_001274737.1 | ||||
NM_001287809.1 | Intron | NP_001274738.1 | ||||
NM_032793.4 | Intron | NP_116182.2 | ||||
XM_005271285.1 | Intron | XP_005271342.1 | ||||
XM_011542312.2 | Intron | XP_011540614.1 |