Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACTAGTTTGCCCAGATTGCCC[A/G]CTAGTAAAGTGGCCTTATTGAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAB2 - TGF-beta activated kinase 1/MAP3K7 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292034.2 | Intron | NP_001278963.1 | ||||
NM_001292035.2 | Intron | NP_001278964.1 | ||||
NM_015093.5 | Intron | NP_055908.1 | ||||
XM_011535633.2 | Intron | XP_011533935.1 | ||||
XM_017010591.1 | Intron | XP_016866080.1 | ||||
XM_017010592.1 | Intron | XP_016866081.1 |