Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGATTTTCTAAGTAATTATTA[C/T]ACCAGAAAGGTCAGGTTAGCCTTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 109675 | |||||||||||||||||||||||
Literature Links: |
ST6GAL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ST6GAL1 - ST6 beta-galactoside alpha-2,6-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003032.2 | Intron | NP_003023.1 | ||||
NM_173216.2 | Intron | NP_775323.1 | ||||
NM_173217.2 | Intron | NP_775324.1 | ||||
XM_005247719.1 | Intron | XP_005247776.1 | ||||
XM_005247720.1 | Intron | XP_005247777.1 | ||||
XM_006713734.1 | Intron | XP_006713797.1 | ||||
XM_011513085.1 | Intron | XP_011511387.1 | ||||
XM_011513086.1 | Intron | XP_011511388.1 | ||||
XM_017007065.1 | Intron | XP_016862554.1 | ||||
XM_017007066.1 | Intron | XP_016862555.1 | ||||
XM_017007067.1 | Intron | XP_016862556.1 | ||||
XM_017007068.1 | Intron | XP_016862557.1 |