Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTGGTTCTTTCAAAGTTATTT[A/C]TGACCTATACTACTGTTTGCCCTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606465 | ||||||||||||||||||||
Literature Links: |
KLF15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLF15 - Kruppel like factor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014079.3 | Intron | NP_054798.1 | ||||
XM_005247400.3 | Intron | XP_005247457.1 | ||||
XM_011512743.2 | Intron | XP_011511045.1 |