Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAATAGTTTGAGAATTATTAAA[A/G]TAAAACTGCTTTTTCAATACAGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607546 | ||||||||||||||||||||
Literature Links: |
CD200R1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD200R1 - CD200 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138806.3 | Intron | NP_620161.1 | ||||
NM_138939.2 | Intron | NP_620385.1 | ||||
NM_138940.2 | Intron | NP_620386.1 | ||||
NM_170780.2 | Intron | NP_740750.1 |