Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCTGGCAGGAGGAATGCTCCAA[G/T]AACGGCAGCACTAAATCTAGTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604862 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD207 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD207 - CD207 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015717.4 | Intron | NP_056532.4 | ||||
XM_011532874.1 | Intron | XP_011531176.1 | ||||
XM_011532875.2 | Intron | XP_011531177.1 | ||||
XM_011532876.2 | Intron | XP_011531178.1 |
CLEC4F - C-type lectin domain family 4 member F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |