Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605638 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIRC6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BIRC6 - baculoviral IAP repeat containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4765 - microRNA 4765 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC27 - tetratricopeptide repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193509.1 | Intron | NP_001180438.1 | ||||
NM_017735.4 | Intron | NP_060205.3 | ||||
XM_005264416.1 | Intron | XP_005264473.1 | ||||
XM_011532958.1 | Intron | XP_011531260.1 |