Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACACACATCCATCACTCAAAG[G/T]CATTGCCTGAATTGACTCATGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606831 MIM: 611148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRC4 - NLR family CARD domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199138.1 | Intron | NP_001186067.1 | ||||
NM_001199139.1 | Intron | NP_001186068.1 | ||||
NM_001302504.1 | Intron | NP_001289433.1 | ||||
NM_021209.4 | Intron | NP_067032.3 | ||||
XM_017004619.1 | Intron | XP_016860108.1 |
SLC30A6 - solute carrier family 30 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |