Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGGGGGGCTTTTTCCAAATTGT[A/G]AAACTATCTTTTATCATGAAATAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TESPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TESPA1 - thymocyte expressed, positive selection associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098815.2 | 7442 | UTR 3 | NP_001092285.1 | |||
NM_001136030.2 | 7442 | UTR 3 | NP_001129502.1 | |||
NM_001261844.1 | 7442 | UTR 3 | NP_001248773.1 | |||
NM_014796.2 | 7442 | UTR 3 | NP_055611.1 | |||
XM_005269247.2 | 7442 | UTR 3 | XP_005269304.1 | |||
XM_006719715.3 | 7442 | UTR 3 | XP_006719778.1 | |||
XM_011539035.2 | 7442 | UTR 3 | XP_011537337.1 | |||
XM_011539036.1 | 7442 | UTR 3 | XP_011537338.1 | |||
XM_011539037.2 | 7442 | UTR 3 | XP_011537339.1 | |||
XM_017020262.1 | 7442 | UTR 3 | XP_016875751.1 | |||
XM_017020263.1 | 7442 | UTR 3 | XP_016875752.1 |