Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGGCCCTATAAAGCTTTTATT[C/T]TCCTATGGCATTGCTTAAGAATCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179530 | ||||||||||||||||||||
Literature Links: |
RAP1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAP1B - RAP1B, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010942.2 | Intron | NP_001010942.1 | ||||
NM_001251917.1 | Intron | NP_001238846.1 | ||||
NM_001251918.1 | Intron | NP_001238847.1 | ||||
NM_001251921.1 | Intron | NP_001238850.1 | ||||
NM_001251922.1 | Intron | NP_001238851.1 | ||||
NM_015646.5 | Intron | NP_056461.1 |
SNORA70G - small nucleolar RNA, H/ACA box 70G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |