Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCAGGGGCACCTCGCAGTACCC[A/C]TGCGTCCAGGTCTACGTGAACAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605223 | ||||||||||||||||||||
Literature Links: |
KCNMB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNMB4 - potassium calcium-activated channel subfamily M regulatory beta subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014505.5 | 764 | Silent Mutation | CCA,CCC | P,P 83 | NP_055320.4 | |
XM_011538188.2 | 764 | Silent Mutation | CCA,CCC | P,P 83 | XP_011536490.1 |