Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCACACCGCCTGAAGCTTCAGC[C/T]ACACTGAAACTTCATAACAAAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604979 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPSF6 - cleavage and polyadenylation specific factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300947.1 | Intron | NP_001287876.1 | ||||
NM_007007.2 | Intron | NP_008938.2 | ||||
XM_005268588.2 | Intron | XP_005268645.1 | ||||
XM_005268590.2 | Intron | XP_005268647.1 |
Set Membership: |
HapMap |