Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGGAGCAAAATTCAATGGTATG[A/G]ATAGAGATTTTAAACAAAACAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYRFL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYRFL - myelin regulatory factor-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182530.2 | Intron | NP_872336.2 | ||||
XM_011539052.2 | Intron | XP_011537354.1 | ||||
XM_011539055.1 | Intron | XP_011537357.1 | ||||
XM_017018967.1 | Intron | XP_016874456.1 | ||||
XM_017018968.1 | Intron | XP_016874457.1 | ||||
XM_017018969.1 | Intron | XP_016874458.1 | ||||
XM_017018970.1 | Intron | XP_016874459.1 | ||||
XM_017018971.1 | Intron | XP_016874460.1 | ||||
XM_017018972.1 | Intron | XP_016874461.1 | ||||
XM_017018973.1 | Intron | XP_016874462.1 | ||||
XM_017018974.1 | Intron | XP_016874463.1 | ||||
XM_017018975.1 | Intron | XP_016874464.1 | ||||
XM_017018976.1 | Intron | XP_016874465.1 |
RAB3IP - RAB3A interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |