Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAACATCACACTCTTGATTTCAT[C/G]GACATAACTGCCAAGAAAAGTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602692 MIM: 612817 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLIPR1 - GLI pathogenesis related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006851.2 | 3062 | Intron | NP_006842.2 | |||
XM_011537775.2 | 3062 | Intron | XP_011536077.1 | |||
XM_017018721.1 | 3062 | Intron | XP_016874210.1 |
KRR1 - KRR1, small subunit processome component homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007043.6 | 3062 | UTR 3 | NP_008974.5 |
Set Membership: |
HapMap |