Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACTCTTTCACCAAGAAGTGTCT[C/T]TTTTAGAAATCTATTCTGCAGATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609147 | ||||||||||||||||||||
Literature Links: |
RIC8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIC8B - RIC8 guanine nucleotide exchange factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268998.3 | Intron | XP_005269055.1 | ||||
XM_011538526.2 | Intron | XP_011536828.1 | ||||
XM_011538527.2 | Intron | XP_011536829.1 | ||||
XM_011538528.2 | Intron | XP_011536830.1 | ||||
XM_011538529.2 | Intron | XP_011536831.1 | ||||
XM_017019548.1 | Intron | XP_016875037.1 | ||||
XM_017019549.1 | Intron | XP_016875038.1 |