Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGTACTCACTGAGCTCCTGCT[C/G]TAGCCCTAGCGGTGTTCTAAGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCP11L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCP11L2 - t-complex 11 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286262.1 | Intron | NP_001273191.1 | ||||
NM_152772.2 | Intron | NP_689985.1 | ||||
XM_005268767.4 | Intron | XP_005268824.1 | ||||
XM_005268768.4 | Intron | XP_005268825.1 | ||||
XM_011538129.2 | Intron | XP_011536431.1 | ||||
XM_011538130.2 | Intron | XP_011536432.1 | ||||
XM_011538131.2 | Intron | XP_011536433.1 | ||||
XM_011538132.2 | Intron | XP_011536434.1 | ||||
XM_017019127.1 | Intron | XP_016874616.1 | ||||
XM_017019128.1 | Intron | XP_016874617.1 | ||||
XM_017019129.1 | Intron | XP_016874618.1 | ||||
XM_017019130.1 | Intron | XP_016874619.1 | ||||
XM_017019131.1 | Intron | XP_016874620.1 | ||||
XM_017019132.1 | Intron | XP_016874621.1 | ||||
XM_017019133.1 | Intron | XP_016874622.1 |
Set Membership: |
HapMap |