Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGGAGCCTCCGGGGGTCAGTGC[C/T]GTGTGCTTTAGGCTGCTAGCAAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613421 MIM: 608047 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCTD10 - potassium channel tetramerization domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE3B - ubiquitin protein ligase E3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270449.1 | Intron | NP_001257378.1 | ||||
NM_001270450.1 | Intron | NP_001257379.1 | ||||
NM_001270451.1 | Intron | NP_001257380.1 | ||||
NM_130466.3 | Intron | NP_569733.2 | ||||
NM_183415.2 | Intron | NP_904324.1 | ||||
XM_005253987.2 | Intron | XP_005254044.1 | ||||
XM_006719681.3 | Intron | XP_006719744.1 | ||||
XM_006719682.2 | Intron | XP_006719745.1 | ||||
XM_011538959.2 | Intron | XP_011537261.1 | ||||
XM_011538961.1 | Intron | XP_011537263.1 | ||||
XM_017020195.1 | Intron | XP_016875684.1 | ||||
XM_017020196.1 | Intron | XP_016875685.1 |
Set Membership: |
HapMap |