Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCTCCTCTAACCACCCCATCT[A/G]AAGTGACCTTCCCTTAAACCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610602 MIM: 191525 MIM: 612492 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALKBH2 - alkB homolog 2, alpha-ketoglutarate dependent dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001655.2 | Intron | NP_001001655.1 | ||||
NM_001145374.1 | Intron | NP_001138846.1 | ||||
NM_001145375.1 | Intron | NP_001138847.1 | ||||
NM_001205179.1 | Intron | NP_001192108.1 | ||||
NM_001205180.1 | Intron | NP_001192109.1 | ||||
XM_005253835.4 | Intron | XP_005253892.1 | ||||
XM_005253836.1 | Intron | XP_005253893.1 |
UNG - uracil DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP30 - ubiquitin specific peptidase 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |