Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCAAAGAATGTTTAATGGTTCA[A/G]AAAAAGGAAATGCTGAAAGCCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608771 | ||||||||||||||||||||
Literature Links: |
MED13L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED13L - mediator complex subunit 13 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015335.4 | Intron | NP_056150.1 | ||||
XM_011538080.2 | Intron | XP_011536382.1 | ||||
XM_011538081.2 | Intron | XP_011536383.1 | ||||
XM_011538082.2 | Intron | XP_011536384.1 | ||||
XM_017019090.1 | Intron | XP_016874579.1 |