Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAATTATGCTTATCTATTTCTGC[C/T]TAAATCATCTATCTATGTGCTACCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
METTL6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
METTL6 - methyltransferase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301790.1 | 2997 | Intron | NP_001288719.1 | |||
NM_001301791.1 | 2997 | Intron | NP_001288720.1 | |||
NM_001301792.1 | 2997 | Intron | NP_001288721.1 | |||
NM_152396.3 | 2997 | Intron | NP_689609.2 | |||
XM_005264867.3 | 2997 | UTR 3 | XP_005264924.1 | |||
XM_005264870.4 | 2997 | Intron | XP_005264927.1 | |||
XM_006712970.3 | 2997 | Intron | XP_006713033.1 | |||
XM_006712972.3 | 2997 | UTR 3 | XP_006713035.1 | |||
XM_011533357.2 | 2997 | Intron | XP_011531659.1 | |||
XM_017005718.1 | 2997 | Intron | XP_016861207.1 | |||
XM_017005719.1 | 2997 | UTR 3 | XP_016861208.1 | |||
XM_017005720.1 | 2997 | Intron | XP_016861209.1 | |||
XM_017005721.1 | 2997 | Intron | XP_016861210.1 | |||
XM_017005722.1 | 2997 | Intron | XP_016861211.1 | |||
XM_017005723.1 | 2997 | UTR 3 | XP_016861212.1 | |||
XM_017005724.1 | 2997 | Intron | XP_016861213.1 |