Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGATGAATAATCTGTGTTACT[A/G]CATCATTCCCATAGGTTATAGTTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600043 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1E1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SULT1E1 - sulfotransferase family 1E member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005420.2 | 1719 | UTR 3 | NP_005411.1 | |||
XM_011532210.2 | 1719 | Intron | XP_011530512.1 |
Set Membership: |
HapMap |