Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606769 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELQ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HELQ - helicase, POLQ-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297755.1 | Intron | NP_001284684.1 | ||||
NM_001297756.1 | Intron | NP_001284685.1 | ||||
NM_001297757.1 | Intron | NP_001284686.1 | ||||
NM_001297758.1 | Intron | NP_001284687.1 | ||||
NM_001297759.1 | Intron | NP_001284688.1 | ||||
NM_133636.3 | Intron | NP_598375.2 | ||||
XM_005262711.1 | Intron | XP_005262768.1 | ||||
XM_005262713.2 | Intron | XP_005262770.1 | ||||
XM_006714076.2 | Intron | XP_006714139.1 | ||||
XM_011531580.2 | Intron | XP_011529882.1 | ||||
XM_017007679.1 | Intron | XP_016863168.1 | ||||
XM_017007680.1 | Intron | XP_016863169.1 | ||||
XM_017007681.1 | Intron | XP_016863170.1 | ||||
XM_017007682.1 | Intron | XP_016863171.1 | ||||
XM_017007683.1 | Intron | XP_016863172.1 |
Set Membership: |
HapMap |