Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCCTGCATTCAAAACTTGGAC[C/T]TGTCATTAATTACCCCTGTGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610151 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
METAP1 - methionyl aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015143.2 | Intron | NP_055958.2 | ||||
XM_011531777.2 | Intron | XP_011530079.1 | ||||
XM_011531778.1 | Intron | XP_011530080.1 | ||||
XM_011531779.1 | Intron | XP_011530081.1 | ||||
XM_017007914.1 | Intron | XP_016863403.1 | ||||
XM_017007915.1 | Intron | XP_016863404.1 | ||||
XM_017007916.1 | Intron | XP_016863405.1 | ||||
XM_017007917.1 | Intron | XP_016863406.1 |
Set Membership: |
HapMap |