Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608009 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK5 - adenylate kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZZZ3 - zinc finger ZZ-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308237.1 | 5274 | Intron | NP_001295166.1 | |||
NM_015534.5 | 5274 | Intron | NP_056349.1 | |||
XM_005270725.3 | 5274 | UTR 3 | XP_005270782.1 | |||
XM_005270726.3 | 5274 | Intron | XP_005270783.1 | |||
XM_005270727.3 | 5274 | UTR 3 | XP_005270784.1 | |||
XM_005270729.4 | 5274 | UTR 3 | XP_005270786.1 | |||
XM_017000970.1 | 5274 | UTR 3 | XP_016856459.1 | |||
XM_017000971.1 | 5274 | Intron | XP_016856460.1 | |||
XM_017000972.1 | 5274 | UTR 3 | XP_016856461.1 |
Set Membership: |
HapMap |