Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATTTTCAAAACATCCAAGCCAC[A/T]CTTTAAAAACGAATTACAAAACAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604425 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LHX8 - LIM homeobox 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A5 - solute carrier family 44 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130058.1 | 2640 | UTR 3 | NP_001123530.1 | |||
NM_001320283.1 | 2640 | Intron | NP_001307212.1 | |||
NM_001320285.1 | 2640 | UTR 3 | NP_001307214.1 | |||
NM_001320287.1 | 2640 | UTR 3 | NP_001307216.1 | |||
NM_152697.4 | 2640 | Intron | NP_689910.2 | |||
XM_006710445.3 | 2640 | UTR 3 | XP_006710508.1 | |||
XM_011540986.2 | 2640 | UTR 3 | XP_011539288.1 | |||
XM_011540987.2 | 2640 | UTR 3 | XP_011539289.1 | |||
XM_017000607.1 | 2640 | UTR 3 | XP_016856096.1 | |||
XM_017000608.1 | 2640 | UTR 3 | XP_016856097.1 | |||
XM_017000609.1 | 2640 | UTR 3 | XP_016856098.1 | |||
XM_017000610.1 | 2640 | UTR 3 | XP_016856099.1 | |||
XM_017000611.1 | 2640 | UTR 3 | XP_016856100.1 |
Set Membership: |
HapMap |