Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGAAACCTTTGTTGAAGGACTA[C/G]TGGGAGAAAGAAGGGGTTTAGGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610133 | ||||||||||||||||||||
Literature Links: |
ST6GALNAC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST6GALNAC3 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160011.1 | Intron | NP_001153483.1 | ||||
NM_152996.2 | Intron | NP_694541.2 | ||||
XM_006710553.3 | Intron | XP_006710616.1 | ||||
XM_017000937.1 | Intron | XP_016856426.1 | ||||
XM_017000938.1 | Intron | XP_016856427.1 | ||||
XM_017000939.1 | Intron | XP_016856428.1 | ||||
XM_017000940.1 | Intron | XP_016856429.1 | ||||
XM_017000941.1 | Intron | XP_016856430.1 | ||||
XM_017000942.1 | Intron | XP_016856431.1 |