Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602105 MIM: 179080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002440.3 | 4 | UTR 5 | NP_002431.2 |
RABGGTB - Rab geranylgeranyltransferase beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45A - small nucleolar RNA, C/D box 45A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45B - small nucleolar RNA, C/D box 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45C - small nucleolar RNA, C/D box 45C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |