Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCGCCCGCTGGCAGGCGACACG[C/T]GGTGAGTGACCAGTGACTCGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601642 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL12RB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL12RB2 - interleukin 12 receptor subunit beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258214.1 | 616 | UTR 5 | NP_001245143.1 | |||
NM_001258215.1 | 616 | UTR 5 | NP_001245144.1 | |||
NM_001258216.1 | 616 | UTR 5 | NP_001245145.1 | |||
NM_001319233.1 | 616 | UTR 5 | NP_001306162.1 | |||
NM_001559.2 | 616 | UTR 5 | NP_001550.1 | |||
XM_005270825.2 | 616 | UTR 5 | XP_005270882.1 | |||
XM_005270827.2 | 616 | UTR 5 | XP_005270884.1 | |||
XM_005270828.3 | 616 | UTR 5 | XP_005270885.1 | |||
XM_006710617.2 | 616 | UTR 5 | XP_006710680.1 | |||
XM_011541383.2 | 616 | Intron | XP_011539685.1 | |||
XM_011541384.2 | 616 | UTR 5 | XP_011539686.1 | |||
XM_017001202.1 | 616 | UTR 5 | XP_016856691.1 | |||
XM_017001203.1 | 616 | UTR 5 | XP_016856692.1 | |||
XM_017001204.1 | 616 | UTR 5 | XP_016856693.1 |
Set Membership: |
HapMap |